missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-sulfamoylbenzoic Acid, TRC

Product Code. 30135435
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135435 5 g 5g
17923585 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135435 Supplier Toronto Research Chemicals Supplier No. TRCC3827005G
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1205-30-7
Molecular Formula C7 H6 Cl N O4 S
Molecular Weight (g/mol) 235.64
Synonym 4-Chloro-3-sulfamoylbenzoic Acid, Benzoic acid, 3-(aminosulfonyl)-4-chloro- (9CI, ACI), 3-(Aminosulfonyl)-4-chlorobenzoic acid (ACI), Benzoic acid, 4-chloro-3-sulfamoyl- (7CI, 8CI), 3-Sulfamoyl-4-chlorobenzoic acid, 3-Sulfamyl-4-chlorobenzoic acid, 4-Chloro-3-(aminosulfonyl)benzoic acid, 4-Chloro-3-sulfamoylbenzoic acid, 4-Chloro-5-sulfamoylbenzoic acid
IUPAC Name 4-chloro-3-sulfamoylbenzoic acid
SMILES NS(=O)(=O)c1cc(ccc1Cl)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H6ClNO4S/c8-5-2-1-4(7(10)11)3-6(5)14(9,12)13/h1-3H,(H,10,11)(H2,9,12,13)
Formula Weight 234.9706
Percent Purity >95%
Chemical Name or Material 4-Chloro-3-sulfamoylbenzoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.