missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10486852
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10486852 1 g 1g
10754003 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10486852 Supplier Thermo Scientific Maybridge Supplier No. MO07002DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 32333-53-2
Molecular Formula C7H3Cl2F3O2S
Molecular Weight (g/mol) 279.054
MDL Number MFCD00024882
InChI Key SSULGNXFUGLULI-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl benzenesulfonyl chloride, 4-chloro-3-trifluoromethyl benzene-1-sulfonyl chloride, 4-chloro-3-trifluoromethyl-benzenesulfonyl chloride, 4-chloro-3-trifluoromethyl benzenesulfonylchloride, 4-chloro-3-trifluoromethyl benzenesulphonyl chloride, 4-chloro-5-trifluoromethyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-chloro-3-trifluoromethyl, ksc222e0j, 4-chloro-3-trifluoromethylbenzenesulfonylchloride, 4-chloro-3-trifluoromethyl benzenesulfonyl chlori
PubChem CID 2780719
IUPAC Name 4-chloro-3-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)C(F)(F)F)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 279.06
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.