missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 15269646 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15269646 5 g 5g
15259646 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15269646 Supplier Thermo Scientific Acros Supplier No. 450310050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 32333-53-2
Molecular Formula C7H3Cl2F3O2S
Molecular Weight (g/mol) 279.06
InChI Key SSULGNXFUGLULI-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl benzenesulfonyl chloride, 4-chloro-3-trifluoromethyl benzene-1-sulfonyl chloride, 4-chloro-3-trifluoromethyl-benzenesulfonyl chloride, 4-chloro-3-trifluoromethyl benzenesulfonylchloride, 4-chloro-3-trifluoromethyl benzenesulphonyl chloride, 4-chloro-5-trifluoromethyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-chloro-3-trifluoromethyl, ksc222e0j, 4-chloro-3-trifluoromethylbenzenesulfonylchloride, 4-chloro-3-trifluoromethyl benzenesulfonyl chlori
PubChem CID 2780719
IUPAC Name 4-chloro-3-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)C(F)(F)F)Cl

Specifications

Melting Point 28.0°C to 30.0°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 279.06
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzenesulfonyl chloride
compliance-icons
Product Identifier
  • 4-Chloro-3-(trifluoromethyl)benzenesulfonyl chloride, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.