missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)benzyl chloride, Thermo Scientific™

Product Code. 10362590
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10362590 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10362590 Supplier Thermo Scientific Maybridge Supplier No. MAY00255DA
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 23131-73-9
Packaging Amber Glass Bottle
Molecular Formula C8H5Cl2F3
Quantity 1 g
Synonym 1-chloro-4-chloromethyl-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl benzyl chloride, 3-trifluoromethyl-4-chlorobenzyl chloride, 3-trifluoromethyl-4-chloro-benzyl chloride, 3-trifluoromethyl-4-chlorobenzyl chloridec, 1-chloro-4-chloromethyl-2-trifluoromethyl-benzene, benzene, 1-chloro-4-chloromethyl-2-trifluoromethyl
InChI Key NREUBTUYADXMHQ-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1CCl)C(F)(F)F)Cl
IUPAC Name 1-chloro-4-(chloromethyl)-2-(trifluoromethyl)benzene
Molecular Weight (g/mol) 229.023
PubChem CID 13094262
Formula Weight 229.03
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.