missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-6-methyl-2-methylthiopyrimidine, 97%, Thermo Scientific™

Product Code. 15327390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15327390 1 g 1g
15337390 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15327390 Supplier Thermo Scientific Acros Supplier No. 453750010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 17119-73-2
Molecular Formula C6H7ClN2S
Molecular Weight (g/mol) 174.65
InChI Key ALMBOXQFPLQVLF-UHFFFAOYSA-N
Synonym 4-chloro-6-methyl-2-methylthio pyrimidine, 4-chloro-6-methyl-2-methylsulfanyl pyrimidine, 4-chloro-6-methyl-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-6-methyl-2-methylthio, 4-chloro-6-methyl-2-methylthiopyrimidine, 2-methylmercapto-4-methyl-6-chloropyrimidine, pubchem21461, 6-chloro-4-methyl-2-methylthiopyrimidine, 4-chloro-6-methyl-2-methylthio-pyrimidine, 2-methylthio-4-methyl-6-chloropyrimidine
PubChem CID 241163
IUPAC Name 4-chloro-6-methyl-2-methylsulfanylpyrimidine
SMILES CC1=CC(=NC(=N1)SC)Cl

Specifications

Melting Point 34.0°C to 37.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 174.65
Percent Purity 97%
Chemical Name or Material 4-Chloro-6-methyl-2-methylthiopyrimidine
compliance-icons
Product Identifier
  • 4-Chloro-6-methyl-2-methylthiopyrimidine, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.