missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-8-fluoroquinoline

Product Code. 11379999
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379999 1 g 1g
11389999 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379999 Supplier Thermo Scientific Alfa Aesar Supplier No. H50541.03
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 63010-72-0 | C9H5ClFN | 181.59 g/mol

4-Chloro-8-fluoroquinoline is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-8-fluoroquinoline is used as an intermediate in organic synthesis.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63010-72-0
Molecular Formula C9H5ClFN
Molecular Weight (g/mol) 181.59
MDL Number MFCD00278785
InChI Key NUIMKHDPMIPYFI-UHFFFAOYSA-N
PubChem CID 2736587
IUPAC Name 4-chloro-8-fluoroquinoline
SMILES FC1=C2N=CC=C(Cl)C2=CC=C1

Specifications

Quantity 1 g
Synonym 4-chloro-8-fluoro-quinoline, 8-fluoro-4-chloroquinoline, quinoline, 4-chloro-8-fluoro, pubchem5886, 4-chloro-8fluoroquinoline, 4-chloro-8-fluoroquinolline, quinoline,4-chloro-8-fluoro, 4-chloranyl-8-fluoranyl-quinoline
Formula Weight 181.6
Chemical Name or Material 4-Chloro-8-fluoroquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.