missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-(4-methoxyphenyl)benzamide, 97%

Product Code. 15451728
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451728 250 mg 250mg
15441728 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15451728 Supplier Thermo Scientific Alfa Aesar Supplier No. H55469.MD
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4018-82-0 | C14H12ClNO2 | 261.705 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 4018-82-0
Molecular Formula C14H12ClNO2
Molecular Weight (g/mol) 261.705
MDL Number MFCD00508611
InChI Key UVGPQLMHIRNFEE-UHFFFAOYSA-N
Synonym 4-chloro-n-4-methoxyphenyl benzamide, n-p-chlorobenzoyl-p-anisidine, 4-chloro-n-4-methoxy-phenyl-benzamide, benzamide, 4-chloro-n-4-methoxyphenyl, cambridge id 5190257, n-4-methoxyphenyl-4-chlorobenzamide, 4-chloro-n-4-methoxyphenyl-benzamide, 4-chloro-n-4-methoxyphenyl benzamide #, 4-chlorobenzenamide, n-4-methoxyphenyl
PubChem CID 221848
IUPAC Name 4-chloro-N-(4-methoxyphenyl)benzamide
SMILES COC1=CC=C(C=C1)NC(=O)C2=CC=C(C=C2)Cl

Tekniske data

Quantity 250 mg
Formula Weight 261.71
Percent Purity 97%
Chemical Name or Material 4-Chloro-N-(4-methoxyphenyl)benzamide

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.