missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroanisole, 99%

Product Code. 10462780 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10462780

Brand: Thermo Scientific Acros 170150250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-12-1 | C7H7ClO | 142.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-12-1
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
InChI Key YRGAYAGBVIXNAQ-UHFFFAOYSA-N
Synonym 4-chloroanisole, benzene, 1-chloro-4-methoxy, p-chloroanisole, p-chlorophenyl methyl ether, anisyl chloride, anisole, p-chloro, p-chloromethoxybenzene, 4-chlorophenol methyl ether, 1-chloro-4-methoxy-benzene, unii-0f18bly08q
PubChem CID 12167
IUPAC Name 1-chloro-4-methoxybenzene
SMILES COC1=CC=C(C=C1)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -18.0°C
Color Colorless to Yellow
Density 1.1600g/mL
Boiling Point 198.0°C to 202.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4OCH3
Refractive Index 1.5340 to 1.5360
Quantity 25 g
Beilstein 06,186
Specific Gravity 1.16
Solubility Information Solubility in water: immiscible.
Formula Weight 142.58
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Chloroanisole, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloroanisole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.