missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzoic acid, 98+%

Product Code. 11452108
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452108 250 g 250g
11442108 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11452108 Supplier Thermo Scientific Alfa Aesar Supplier No. A15135.30
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 74-11-3 | C7H5ClO2 | 156.565 g/mol

4-Chlorobenzoic acid is used to determine the diversity of 2-, 3- and 4-chlorobenzoate degraders in two pristine soils with similar physical and chemical characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzoic acid is used to determine the diversity of 2-, 3- and 4-chlorobenzoate degraders in two pristine soils with similar physical and chemical characteristics.

Solubility
Soluble in water(0.08g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-11-3
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.565
MDL Number MFCD00002531
InChI Key XRHGYUZYPHTUJZ-UHFFFAOYSA-N
Synonym p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italian
PubChem CID 6318
ChEBI CHEBI:30747
IUPAC Name 4-chlorobenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)Cl

Specifications

Melting Point 239°C to 243°C
Density 1.54
Boiling Point 275°C
Flash Point 238°C (460°F)
Odor Odorless
Quantity 250 g
Beilstein 907196
Merck Index 14,2125
Solubility Information Soluble in water(0.08g/L).
Formula Weight 156.57
Percent Purity ≥98%
Chemical Name or Material 4-Chlorobenzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.