missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzophenone, 99%

Product Code. 10192560 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10192560 5 g Glass bottle 5g
10636292 100 g Plastic bottle 100g
10244180 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10192560 Supplier Thermo Scientific Acros Supplier No. 108690050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 134-85-0 | C13H9ClO | 216.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-85-0
MDL Number MFCD00000622
InChI Key UGVRJVHOJNYEHR-UHFFFAOYSA-N
Synonym 4-chlorobenzophenone, 4-chlorophenyl phenyl methanone, p-chlorobenzophenone, methanone, 4-chlorophenyl phenyl, benzophenone, 4-chloro, p-cbp, unii-wih1iz728u, 4-chloro benzophenone, 4-chlorophenyl-phenylmethanone, wih1iz728u
PubChem CID 8653
IUPAC Name (4-chlorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 75°C to 77°C
Color White
Boiling Point 330°C to 332°C
Flash Point 143°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C13H9ClO
Linear Formula ClC6H4COC6H5
Quantity 5 g
Beilstein 07, 419
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acetone,benzene,ether,hot alcohol,and carbon tetrachloride
Molecular Weight (g/mol) 216.67
Formula Weight 216.67
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Chlorobenzophenone, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chlorobenzophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.