missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzophenone, 99%

Product Code. 11488047
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488047 100 g 100g
11498047 500 g 500g
11408057 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488047 Supplier Thermo Scientific Alfa Aesar Supplier No. A17458.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 134-85-0 | C13H9ClO | 216.664 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-85-0
Molecular Formula C13H9ClO
Molecular Weight (g/mol) 216.664
MDL Number MFCD00000622
InChI Key UGVRJVHOJNYEHR-UHFFFAOYSA-N
Synonym 4-chlorobenzophenone, 4-chlorophenyl phenyl methanone, p-chlorobenzophenone, methanone, 4-chlorophenyl phenyl, benzophenone, 4-chloro, p-cbp, unii-wih1iz728u, 4-chloro benzophenone, 4-chlorophenyl-phenylmethanone, wih1iz728u
PubChem CID 8653
IUPAC Name (4-chlorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)Cl

Specifications

Melting Point 75°C to 77°C
Boiling Point 332°C
Flash Point 143°C (289°F)
Quantity 100 g
Beilstein 512043
Formula Weight 216.67
Percent Purity 99%
Chemical Name or Material 4-Chlorobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.