missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzotrifluoride, 98+%

Product Code. 11462138
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462138 2500 g 2500g
11442138 100 g 100g
11452138 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462138 Supplier Thermo Scientific Alfa Aesar Supplier No. A15154.0E
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 98-56-6 | C7H4ClF3 | 180.554 g/mol

4-Chlorobenzotrifluoride acts as an important intermediate for dyes. It is used as an active component of polyurethane finishes. It is also used in adhesives, pigments, sealant chemicals, paints and coatings. Further, it is used as an environmental friendly solvent for cleaning or degreasing and dielectric fluid. It has a high capacity to dissolve inks, which is used in the printing industry. In addition to this, it is also used as xylene replacement in cleaners, thinners and other aromatic hydrocarbon blends.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzotrifluoride acts as an important intermediate for dyes. It is used as an active component of polyurethane finishes. It is also used in adhesives, pigments, sealant chemicals, paints and coatings. Further, it is used as an environmental friendly solvent for cleaning or degreasing and dielectric fluid. It has a high capacity to dissolve inks, which is used in the printing industry. In addition to this, it is also used as xylene replacement in cleaners, thinners and other aromatic hydrocarbon blends.

Solubility
Miscible with chloroform. Slightly miscible with water.

Notes
Store in a cool place. Light sensitive. Incompatible with sodium dimethyl sulfonate, strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-56-6
Molecular Formula C7H4ClF3
Molecular Weight (g/mol) 180.554
MDL Number MFCD00000627
InChI Key QULYNCCPRWKEMF-UHFFFAOYSA-N
Synonym 4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene
PubChem CID 7394
IUPAC Name 1-chloro-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)Cl

Specifications

Melting Point -36°C
Density 1.346
Boiling Point 136°C to 138°C
Flash Point 47°C (116°F)
Refractive Index 1.446
Quantity 2500 g
UN Number UN2234
Beilstein 510203
Merck Index 14,2126
Solubility Information Miscible with chloroform. Slightly miscible with water.
Formula Weight 180.56
Percent Purity ≥98%
Chemical Name or Material 4-Chlorobenzotrifluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.