missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzotrifluoride, TRC

Product Code. 30368080
Change view
Click to view available options
Quantity:
1 g
500 g
Unit Size:
1g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368080 500 g 500g
30367648 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30368080 Supplier Toronto Research Chemicals Supplier No. TRCC364525500G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-56-6
Molecular Formula C7 H4 Cl F3
Molecular Weight (g/mol) 180.55
Synonym Toluene, p-chloro-α,α,α-trifluoro- (6CI,7CI,8CI), 1-Chloro-4-(trifluoromethyl)benzene, (p-Chlorophenyl)trifluoromethane, 4-(Trifluoromethyl)chlorobenzene, 4-(Trifluoromethyl)phenyl chloride, 4-Chloro(trifluoromethyl)benzene, 4-Chloro-1-trifluoromethylbenzene, 4-Chloro-α,α,α-benzotrifluoride, 4-Chloro-α,α,α-trifluorotoluene, 4-Chlorobenzotrifluoride, 4-Chlorobenzyltrifluoride, 4-Trifluoromethyl-1-chlorobenzene, NSC 10309, Oxol 100, Oxsol 100, Oxsul 100, p-(Trifluoromethyl)chlorobenzene, p-Chloro(trifluoromethyl)benzene, p-Chloro-α,α,α-trifluorotoluene, p-Chlorobenzotrifluoride, p-Trifluoromethylphenyl chloride
IUPAC Name 1-chloro-4-(trifluoromethyl)benzene
SMILES FC(F)(F)c1ccc(Cl)cc1

Specifications

Quantity 500 g
InChI Formula InChI=1S/C7H4ClF3/c8-6-3-1-5(2-4-6)7(9,10)11/h1-4H
Formula Weight 179.9954
Chemical Name or Material 4-Chloro-alpha,alpha,alpha-trifluorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.