missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl Chloride (~90%), TRC

Product Code. 30138658
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138658 100 g 100g
30005644 50 g 50g
30011261 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138658 Supplier Toronto Research Chemicals Supplier No. TRCC364730100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-83-6
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.03
Synonym (4-Chlorophenyl)methyl Chloride, 1-(Chloromethyl)-4-chlorobenzene, 1-Chloro-4-(chloromethyl)benzene, 4-Chloro-1-(chloromethyl)benzene, 4-Chlorobenzyl Chloride, 4-Chloromethyl chlorobenzene, NSC 34032, p,alpha-Dichlorotoluene, p-Chlorobenzyl Chloride, p-Chlorophenylmethyl Chloride, alpha,4-Dichlorotoluene, alpha,p-Dichlorotoluene;
IUPAC Name 1-chloro-4-(chloromethyl)benzene
SMILES ClCc1ccc(Cl)cc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C7H6Cl2/c8-5-6-1-3-7(9)4-2-6/h1-4H,5H2
Formula Weight 159.9847
Chemical Name or Material 4-Chlorobenzyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.