missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl chloride, 98+%

Product Code. p-7022088
Click to view available options
Quantity:
25 g
250 g
1000 g
5000 g
Unit Size:
1000g
250g
25g
5000g
This item is not returnable. View return policy

Product Code. 11364749

Brand: Thermo Scientific Alfa Aesar A14696.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-83-6 | C7H6Cl2 | 161.025 g/mol

4-Chlorobenzyl chloride was the test compound for determining rat hepatocellular thiols by HPLC. As a reagent used in derivatization of, for example, a calix[4]arene and a tryptamine. It is widely used as an intermediate for manufacturing organic compounds like benzyl alcohol and benzyl cyanide. Also used in fuel as a gum inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzyl chloride was the test compound for determining rat hepatocellular thiols by HPLC. As a reagent used in derivatization of, for example, a calix[4]arene and a tryptamine. It is widely used as an intermediate for manufacturing organic compounds like benzyl alcohol and benzyl cyanide. Also used in fuel as a gum inhibitor.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-83-6
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000914
InChI Key JQZAEUFPPSRDOP-UHFFFAOYSA-N
Synonym 4-chlorobenzyl chloride, 1-chloro-4-chloromethyl benzene, p-chlorobenzyl chloride, benzene, 1-chloro-4-chloromethyl, alpha,4-dichlorotoluene, 4-chlorobenzylchloride, 4-chlorophenyl methyl chloride, 1-chloro-4-chloromethyl-benzene, alpha,p-dichlorotoluene, toluene, p,alpha-dichloro
PubChem CID 7723
IUPAC Name 1-chloro-4-(chloromethyl)benzene
SMILES C1=CC(=CC=C1CCl)Cl

Specifications

Melting Point 26°C to 30°C
Density 1.275
Boiling Point 220°C to 221°C
Flash Point 107°C (225°F)
Odor Pungent
Refractive Index 1.5575
Quantity 25 g
UN Number UN3427
Beilstein 471558
Solubility Information Insoluble in water.
Formula Weight 161.03
Percent Purity ≥98%
Chemical Name or Material 4-Chlorobenzyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.