missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl chloride, 99+%

Product Code. p-85677 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
1 kg
Unit Size:
1kg
250g
5g
This item is not returnable. View return policy

Product Code. 10697112

Brand: Thermo Scientific Acros 150240050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-83-6 | C7H6Cl2 | 161.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-83-6
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.03
MDL Number MFCD00000914
InChI Key JQZAEUFPPSRDOP-UHFFFAOYSA-N
Synonym 4-chlorobenzyl chloride, 1-chloro-4-chloromethyl benzene, p-chlorobenzyl chloride, benzene, 1-chloro-4-chloromethyl, alpha,4-dichlorotoluene, 4-chlorobenzylchloride, 4-chlorophenyl methyl chloride, 1-chloro-4-chloromethyl-benzene, alpha,p-dichlorotoluene, toluene, p,alpha-dichloro
PubChem CID 7723
IUPAC Name 1-chloro-4-(chloromethyl)benzene
SMILES C1=CC(=CC=C1CCl)Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 27°C to 28°C
Color Colorless to White
Boiling Point 221°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 0.5% max. p-Chlorobenzal chloride (GC)
Packaging Glass bottle
Linear Formula ClC6H4CH2Cl
Quantity 5 g
Beilstein 05, 308
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 161.03
Percent Purity 99+%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4-Chlorobenzyl chloride, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.