missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)benzoic acid, 98%

Product Code. 10424895 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 10424895

Brand: Thermo Scientific Acros 392890500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1642-81-5 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1642-81-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 202°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H7ClO2
Linear Formula ClCH2C6H4COOH
MDL Number MFCD00002568
Quantity 50 g
Synonym 4-chloromethyl benzoic acid, alpha-chloro-p-toluic acid, benzoic acid, 4-chloromethyl, 4-carboxybenzyl chloride, 4-chloromethyl-benzoic acid, alpha-chloro-p-toluylic acid, .alpha.-chloro-p-toluylic acid, pubchem15446, alpha-chloroparatoluic acid
InChI Key OITNBJHJJGMFBN-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CCl)C(=O)O
IUPAC Name 4-(chloromethyl)benzoic acid
Molecular Weight (g/mol) 170.59
PubChem CID 74234
Formula Weight 170.59
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Chloromethyl)benzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Chloromethyl)benzoic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.