missing translation for 'onlineSavingsMsg'
Learn More

4-Chloromethylbenzonitrile, 98%

Product Code. 10793435 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10793435

Brand: Thermo Scientific Acros 391150500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 874-86-2 | C8H6ClN | 151.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-86-2
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.59
MDL Number MFCD00019761
InChI Key LOQLDQJTSMKBJU-UHFFFAOYSA-N
Synonym 4-chloromethyl benzonitrile, 4-cyanobenzyl chloride, 4-chloromethyl-benzonitrile, benzonitrile, 4-chloromethyl, ccris 5103, p-cyanobenzyl chloride, 4-chloromethyl benzenecarbonitrile, 4-cyanobenzylchloride, pubchem7377, alp.-chloro-p-tolunitrile
PubChem CID 70127
IUPAC Name 4-(chloromethyl)benzonitrile
SMILES C1=CC(=CC=C1CCl)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 77°C to 80°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2C6H4CN
Quantity 50 g
Formula Weight 151.59
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloromethylbenzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloromethylbenzonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.