missing translation for 'onlineSavingsMsg'
Learn More

4-chlorophenol, 99+%

Product Code. 10504304 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
2.5kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10504304 2.5 kg Glass bottle 2.5kg
10296090 100 g Glass bottle 100g
10022350 500 g Glass bottle 500g
10311194 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10504304 Supplier Thermo Scientific Acros Supplier No. 181000025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-48-9 | C6H5ClO | 128.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-48-9
Molecular Formula C6H5ClO
Molecular Weight (g/mol) 128.56
MDL Number MFCD00002318
InChI Key WXNZTHHGJRFXKQ-UHFFFAOYSA-N
Synonym p-chlorophenol, parachlorophenol, phenol, 4-chloro, 4-hydroxychlorobenzene, phenol, p-chloro, applied 3-78, 4-monochlorophenol, p-chlorophenic acid, p-chlorfenol, 4-chloro-1-hydroxybenzene
PubChem CID 4684
ChEBI CHEBI:28078
IUPAC Name 4-chlorophenol
SMILES C1=CC(=CC=C1O)Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 41.0°C to 45.0°C
Color White to Yellow
Density 1.2600g/mL
Boiling Point 220.0°C
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4OH
Quantity 2.5 kg
Beilstein 06,186
Merck Index 15,2157
Specific Gravity 1.26
Solubility Information Solubility in water: 2.7g/100 mL (20°C). Other solubilities: soluble in alcohol,ether,aromatic solvents,acetone,chloroform,glycerin and sodium hydroxide
Viscosity 4.99 mPa.s (50°C)
Formula Weight 128.56
Percent Purity 99+%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4-Chlorophenol
Show More Show Less

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.