missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorophenyl)[4-(1-methylethoxy)phenyl]methanone(Fenofibrate Impurity), TRC

Product Code. 30135346
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135346 100 mg 100mg
30138096 250 mg 250mg
30007129 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135346 Supplier Toronto Research Chemicals Supplier No. TRCC375150100MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154356-96-4
Molecular Formula C16 H15 Cl O2
Molecular Weight (g/mol) 274.74
Synonym (4-Chlorophenyl)[4-(1-methylethoxy)phenyl]methanone, Fenofibrate Imp. F (EP)
IUPAC Name (4-chlorophenyl)-(4-propan-2-yloxyphenyl)methanone
SMILES CC(C)Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H15ClO2/c1-11(2)19-15-9-5-13(6-10-15)16(18)12-3-7-14(17)8-4-12/h3-11H,1-2H3
Formula Weight 274.0761
Percent Purity >95%
Chemical Name or Material (4-Chlorophenyl)[4-(1-methylethoxy)phenyl]methanone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.