missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylmagnesium bromide, 1M solution in THF/toluene, AcroSeal™

Product Code. 10699414 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10699414 100 mL AcroSeal™ Glass bottle 100mL
10566753 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10699414 Supplier Thermo Scientific Acros Supplier No. 377251000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 873-77-8 | C6H4BrClMg | 215.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-77-8 , 109-99-9 , 108-88-3
Molecular Formula C6H4BrClMg
Molecular Weight (g/mol) 215.76
MDL Number MFCD00009926
InChI Key CDEMHJCJMMOFMB-UHFFFAOYSA-M
Synonym 4-chlorophenylmagnesium bromide, bromo p-chlorophenyl magnesium, bromo 4-chlorophenyl magnesium, p-chlorophenylmagnesium bromide, 4-chlorophenyl magnesium bromide, pubchem24069, 4-chlorophenylmagnesiumbromide, 4-chlorphenylmagnesium bromide, 4-chloro phenylmagnesiumbromide, 4-chlorophenyhnagnesium bromide, grignard reagent
PubChem CID 101888
SMILES ClC1=CC=C([Mg]Br)C=C1

Specifications

Color Yellowish-green solution
CAS Min % 21.58, 76.27
CAS Max % 23.73, 78.42
Linear Formula Cl(C6H6)MgBr
Concentration 1M, exact strength on the certificate of analysis
Density 1.0000g/mL
Solubility Information Solubility in water: vigorous reaction.
IUPAC Name bromo(4-chlorophenyl)magnesium
Formula Weight 215.76
Packaging AcroSeal™ Glass bottle
Specific Gravity 1
Quantity 100 mL
Flash Point −21°C
Chemical Name or Material 4-Chlorophenylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Chlorophenylmagnesium bromide, 0.9M solution in THF/toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.