missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylsulfonylacetone, 98+%, Thermo Scientific™

Product Code. 10311853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10311853 5 g 5g
10291313 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10311853 Supplier Thermo Scientific Acros Supplier No. 372110050
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5000-48-6
Molecular Formula C9H9ClO3S
Molecular Weight (g/mol) 232.68
MDL Number MFCD00018663
InChI Key BRDBHPZILGTBFY-UHFFFAOYSA-N
Synonym 4-chlorophenylsulfonylacetone, 1-4-chlorophenyl sulfonyl acetone, 1-4-chlorophenyl sulfonyl propan-2-one, 4-chlorobenzenesulphonylacetone, p-chlorophenylsulfonylacetone, 1-4-chlorobenzenesulfonyl propan-2-one, 4-chlorophenylsulfonyl acetone, 1-4-chlorophenyl sulfonylpropan-2-one, maybridge1_004685
PubChem CID 521269
IUPAC Name 1-(4-chlorophenyl)sulfonylpropan-2-one
SMILES CC(=O)CS(=O)(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 89°C to 91°C
Assay Percent Range 98% min. (GC)
Packaging Glass Bottle
Linear Formula CCl6H4SO2CH2COCH3
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 232.69
Percent Purity 98+%
Chemical Name or Material 4-Chlorophenylsulfonylacetone
compliance-icons
Product Identifier
  • 4-Chlorophenylsulfonylacetone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.