missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorophenylthio)acetic acid, 98%

Product Code. 11392586
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392586 10 g 10g
11302596 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392586 Supplier Thermo Scientific Alfa Aesar Supplier No. L08583.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3405-88-7 | C8H7ClO2S | 202.652 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3405-88-7
Molecular Formula C8H7ClO2S
Molecular Weight (g/mol) 202.652
MDL Number MFCD00021762
InChI Key YPKLXLYGMAWXDO-UHFFFAOYSA-N
Synonym 4-chlorophenylthio acetic acid, 2-4-chlorophenyl thio acetic acid, 4-chlorophenyl thio acetic acid, p-chlorophenylmercaptoacetic acid, 2-4-chlorophenyl sulfanyl acetic acid, 2-p-chlorophenylthio acetic acid, p-chlorophenylthioacetic acid, 4-chlorophenyl sulfanyl acetic acid, acetic acid, p-chlorophenyl thio, acetic acid, 4-chlorophenyl thio
PubChem CID 55471
IUPAC Name 2-(4-chlorophenyl)sulfanylacetic acid
SMILES C1=CC(=CC=C1SCC(=O)O)Cl

Specifications

Melting Point 104°C to 107°C
Quantity 10 g
Beilstein 1871569
Formula Weight 202.66
Percent Purity 98%
Chemical Name or Material (4-Chlorophenylthio)acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.