missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothioanisole, 98%, Thermo Scientific™

Product Code. 10367711 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Pakningsstørrelse
10367711 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10367711 Leverandør Thermo Scientific Leverandørnr. 331350050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Tekniske data

CAS 123-09-1
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2200g/mL
Boiling Point 168°C to 169°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H7ClS
Linear Formula ClC6H4SCH3
Refractive Index 1.5987 to 1.6007
MDL Number MFCD00013643
Quantity 5 g
Specific Gravity 1.22
Synonym 4-chlorothioanisole, 4-chlorophenyl methyl sulfide, p-chlorothioanisole, 1-chloro-4-methylthio benzene, p-chlorophenyl methyl sulfide, methyl 4-chlorophenyl sulfide, benzene, 1-chloro-4-methylthio, p-chlophenyl methyl sulfide, 1-chloro-4-methylsulfanyl benzene, 4-chloro thioanisole
InChI Key KIQQUVJOLVCZKG-UHFFFAOYSA-N
SMILES CSC1=CC=C(C=C1)Cl
IUPAC Name 1-chloro-4-methylsulfanylbenzene
Molecular Weight (g/mol) 158.65
PubChem CID 31243
Formula Weight 158.65
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 4-Chlorothioanisole, 98%
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.