missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothiophenol, 98%

Product Code. 10427370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10427370 5 g Glass bottle 5g
10407560 100 g Glass bottle 100g
10327520 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10427370 Supplier Thermo Scientific Acros Supplier No. 154760050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106-54-7 | C6H5ClS | 144.616 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-54-7
MDL Number MFCD00004847
InChI Key VZXOZSQDJJNBRC-UHFFFAOYSA-N
Synonym 4-chlorothiophenol, p-chlorothiophenol, benzenethiol, 4-chloro, p-chlorobenzenethiol, p-chlorthiofenol, p-chlorophenylmercaptan, benzenethiol, p-chloro, p-mercaptochlorobenzene, 4-chlorophenylmercaptan, 1-chloro-4-mercaptobenzene
PubChem CID 7815
IUPAC Name 4-chlorobenzenethiol
SMILES C1=CC(=CC=C1S)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 49°C to 53°C
Color White
Boiling Point 205°C to 207°C
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H5ClS
Linear Formula ClC6H4SH
Quantity 5 g
Beilstein 06, 326
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most common organic solvents
Molecular Weight (g/mol) 144.62
Formula Weight 144.62
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4-Chlorothiophenol, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Chlorothiophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.