missing translation for 'onlineSavingsMsg'
Learn More

4-Cumylphenol, 97%

Product Code. 11401027
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11401027 250 g 250g
11374739 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11401027 Supplier Thermo Scientific Alfa Aesar Supplier No. B23410.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 599-64-4 | C15H16O | 212.292 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-64-4
Molecular Formula C15H16O
Molecular Weight (g/mol) 212.292
MDL Number MFCD00002365
InChI Key QBDSZLJBMIMQRS-UHFFFAOYSA-N
Synonym 4-cumylphenol, p-cumylphenol, 4-2-phenylpropan-2-yl phenol, p-cumyl phenol, phenol, 4-1-methyl-1-phenylethyl, 4-1-methyl-1-phenylethyl phenol, 4-alpha-cumylphenol, 4-dimethylphenylmethyl phenol, 4-hydroxydiphenyldimethylmethane, 2-phenyl-2-4-hydroxyphenyl propane
PubChem CID 11742
ChEBI CHEBI:35092
IUPAC Name 4-(2-phenylpropan-2-yl)phenol
SMILES CC(C)(C1=CC=CC=C1)C2=CC=C(C=C2)O

Specifications

Melting Point 70°C to 73°C
Density 1.115
Boiling Point 189°C to 191°C (10 mmHg)
Flash Point 160°C (320°F)
Quantity 250 g
Beilstein 1870517
Formula Weight 212.29
Percent Purity 97%
Chemical Name or Material 4-Cumylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.