missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-1-butyne, 97%

Product Code. 15325028 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15325028

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19596-07-7 | C5H5N | 79.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19596-07-7
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.1
InChI Key VMUWIDHKAIGONP-UHFFFAOYSA-N
Synonym 4-cyano-1-butyne, 4-pentynenitrile, 1-cyano-3-butyne
PubChem CID 140560
IUPAC Name pent-4-ynenitrile
SMILES C#CCCC#N

Specifications

Density 0.9063g/mL
Boiling Point 79.0°C (39.0 mmHg)
Flash Point 65.5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula NCCH2CH2CCH
Quantity 1 g
Specific Gravity 0.9063
Formula Weight 79.1
Percent Purity 97%
Chemical Name or Material 4-Cyano-1-butyne
Show More Show Less
compliance-icons
Product Identifier
  • 4-Cyano-1-butyne
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.