missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-n-hexylbiphenyl, 97%, Thermo Scientific™

Product Code. 15472178
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15472178

Brand: Thermo Scientific Alfa Aesar H55825.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Cyano-4′-n-hexylbiphenyl, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a liquid crystal monomer.

Solubility
Sparingly Soluble in water (3.2E-5 g/L) (25°C).

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41122-70-7
Molecular Formula C19H21N
Molecular Weight (g/mol) 263.384
MDL Number MFCD00075144
InChI Key VADSDVGLFDVIMG-UHFFFAOYSA-N
Synonym 4-hexyl-4'-cyanobiphenyl, 4-cyano-4'-hexylbiphenyl, 4'-hexyl-4-biphenylcarbonitrile, 4-n-hexyl-4'-cyanobiphenyl, 4'-hexyl-1,1'-biphenyl-4-carbonitrile, 4'-hexylbiphenyl-4-carbonitrile, ccris 6044, 1,1'-biphenyl-4-carbonitrile, 4'-hexyl, 4-4-hexylphenyl benzonitrile
PubChem CID 38764
IUPAC Name 4-(4-hexylphenyl)benzonitrile
SMILES CCCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Flash Point 113°C (235°F)
Quantity 1 g
Solubility Information Sparingly Soluble in water (3.2E-5g/L) (25°C).
Formula Weight 263.38
Percent Purity 97%
Chemical Name or Material 4-Cyano-4'-n-hexylbiphenyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.