missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzaldehyde, 98+%

Product Code. 11442727
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442727 100 g 100g
11422727 5 g 5g
11432727 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442727 Supplier Thermo Scientific Alfa Aesar Supplier No. A14914.22
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 105-07-7 | C8H5NO | 131.13 g/mol

4-Cyanobenzaldehyde, is used as an intermediate for organic synthesis.It is also used as an intermediate in the pharmaceutical and research department.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanobenzaldehyde, is used as an intermediate for organic synthesis.It is also used as an intermediate in the pharmaceutical and research department.

Solubility
It is soluble in water, ethanol, ether and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong acids, Strong bases, Strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-07-7
Molecular Formula C8H5NO
Molecular Weight (g/mol) 131.13
MDL Number MFCD00003376
InChI Key WZWIQYMTQZCSKI-UHFFFAOYSA-N
Synonym 4-cyanobenzaldehyde, benzonitrile, 4-formyl, p-cyanobenzaldehyde, p-formylbenzonitrile, benzaldehyde, p-cyano, benzonitrile, p-formyl, usaf kf-1, terephthalaldehydonitrile, p-cyanobenzenecarboxaldehyde, 4-formyl-benzonitrile
PubChem CID 66042
IUPAC Name 4-formylbenzonitrile
SMILES O=CC1=CC=C(C=C1)C#N

Specifications

Melting Point 99°C to 103°C
Boiling Point 133°C (12 mmHg)
Odor Characteristic
Quantity 100 g
Beilstein 606473
Sensitivity Air Sensitive
Solubility Information It is soluble in water,ethanol,ether and chloroform.
Formula Weight 131.13
Percent Purity ≥98%
Chemical Name or Material 4-Cyanobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.