missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobiphenyl, 95%

Product Code. 11336889
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336889 10 g 10g
11346889 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336889 Supplier Thermo Scientific Alfa Aesar Supplier No. H26791.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2920-38-9 | C13H9N | 179.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2920-38-9
Molecular Formula C13H9N
Molecular Weight (g/mol) 179.22
MDL Number MFCD00001821
InChI Key BPMBNLJJRKCCRT-UHFFFAOYSA-N
Synonym 4-cyanobiphenyl, 1,1'-biphenyl-4-carbonitrile, 4-biphenylcarbonitrile, p-phenylbenzonitrile, p-cyanobiphenyl, biphenyl-4-carbonitrile, 4-phenylbenzenecarbonitrile, 4-biphenylcarboxylic acid nitrile, 4-cyanodiphenyl
PubChem CID 18021
IUPAC Name 4-phenylbenzonitrile
SMILES N#CC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 85°C to 87°C
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN3439
Beilstein 1907924
Formula Weight 179.22
Percent Purity 95%
Chemical Name or Material 4-Cyanobiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.