missing translation for 'onlineSavingsMsg'
Learn More

4-(Cyanomethyl)phenylboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 11976071 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11976071 1 g 1g
11986071 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11976071

Brand: Thermo Scientific Acros 444740010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138500-86-4
Molecular Formula C14H18BNO2
Molecular Weight (g/mol) 243.11
MDL Number MFCD02179459
InChI Key URWMFRYGXSHPRV-UHFFFAOYSA-N
Synonym 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethyl benzeneboronic acid pinacol ester, 4-cyanomethyl phenylboronic acid pinacol ester, 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethylphenylboronic acid pinacol ester, 4-cyanomethylphenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetonitrile
PubChem CID 3842922
IUPAC Name 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(CC#N)C=C1

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 243.12
Chemical Name or Material 4-(Cyanomethyl)phenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 4-(Cyanomethyl)phenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.