missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenylhydrazine hydrochloride, 97+%

Product Code. 10380464 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10380464 1 g Glass bottle 1g
10248943 5 g Glass bottle 5g
10627264 25 g Glass bottle 25g
13428538 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10380464 Supplier Thermo Scientific Acros Supplier No. 392300010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2863-98-1 | C7H7N3 | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2863-98-1
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00673994
InChI Key DZUUSHCOMPROCJ-UHFFFAOYSA-N
Synonym 4-cyanophenylhydrazine hydrochloride, 4-hydrazinylbenzonitrile hydrochloride, 4-hydrazinobenzonitrile hydrochloride, 4-cyanophenylhydrazine hcl, benzonitrile, 4-hydrazino-, monohydrochloride, 4-hydrazino-benzonitrile; hydrochloride, pubchem13684, 4-cyanophenylhydrazine-hcl, acmc-1co7p, ksc205o1j
PubChem CID 16212962
SMILES NNC1=CC=C(C=C1)C#N

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 249.0°C
Color Brown to Orange
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Linear Formula NCC6H4NHNH2·HCl
Quantity 1 g
Solubility Information Solubility in water: soluble
IUPAC Name 4-hydrazinylbenzonitrile;hydrochloride
Formula Weight 169.61
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material 4-Cyanophenylhydrazine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Cyanophenylhydrazine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.