missing translation for 'onlineSavingsMsg'
Learn More

4-Dechloro-4-(3-chlorophenyl) Haloperidol Decanoate, TRC

Product Code. 30007966
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007966 10 mg 10mg
30012662 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007966 Supplier Toronto Research Chemicals Supplier No. TRCD22648010MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1797131-50-0
Molecular Formula C37 H45 Cl F N O3
Molecular Weight (g/mol) 606.21
Synonym Haloperidol Decanoate EP Impurity F
IUPAC Name [4-[4-(3-chlorophenyl)phenyl]-1-[4-(4-fluorophenyl)-4-oxo-butyl]-4-piperidyl] decanoate
SMILES CCCCCCCCCC(=O)OC1(CCN(CCCC(=O)c2ccc(F)cc2)CC1)c3ccc(cc3)c4cccc(Cl)c4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C37H45ClFNO3/c1-2-3-4-5-6-7-8-14-36(42)43-37(32-19-15-29(16-20-32)31-11-9-12-33(38)28-31)23-26-40(27-24-37)25-10-13-35(41)30-17-21-34(39)22-18-30/h9,11-12,15-22,28H,2-8,10,13-14,23-27H2,1H3
Formula Weight 605.3072
Chemical Name or Material 4-Dechloro-4-(3-chlorophenyl) Haloperidol Decanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.