missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-(Dicyanomethylene)-2-methyl-6-(p-dimethylaminostyryl)-4H-pyran, 96+%

Product Code. 10617371 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10617371 250 mg 250mg
10186593 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10617371 Supplier Thermo Scientific Chemicals Supplier No. 407072500
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 51325-91-8 | C19H17N3O | 303.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51325-91-8
Molecular Formula C19H17N3O
Molecular Weight (g/mol) 303.35
MDL Number MFCD00051341
InChI Key YLYPIBBGWLKELC-RMKNXTFCSA-N
Synonym unii-9s1wxc1zgn, dcm dye, dcm1, 9s1wxc1zgn, 4-dicyanomethylene-2-methyl-6-4-dimethylaminostyryl-4h-pyran, 4-dicyanomethylene-2-methyl-6-4-dimethylamino styryl-4h-pyran, e-2-2-4-dimethylamino styryl-6-methyl-4h-pyran-4-ylidene malononitrile, 2-2-4-dimethylamino phenyl ethenyl-6-methyl-4h-pyran-4-ylidene propanedinitrile, 2-2-4-dimethylamino styryl-6-methyl-4h-pyran-4-ylidene malononitrile
PubChem CID 688222
IUPAC Name 2-[2-[(E)-2-[4-(dimethylamino)phenyl]ethenyl]-6-methylpyran-4-ylidene]propanedinitrile
SMILES CC1=CC(=C(C#N)C#N)C=C(O1)C=CC2=CC=C(C=C2)N(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 212.0°C
Color Red
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 2841
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene,toluene,chloroform
Formula Weight 303.35
Percent Purity 96+%
Grade Extra Pure
Physical Form Crystalline Needles or Powder
Chemical Name or Material 4-(Dicyanomethylene)-2-methyl-6-(p-dimethylaminostyryl)-4H-pyran, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-(Dicyanomethylene)-2-methyl-6-(p-dimethylaminostyryl)-4H-pyran
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 4-(Dicyanomethylene)-2-methyl-6-(p-dimethylaminostyryl)-4H-pyran

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.