missing translation for 'onlineSavingsMsg'
Learn More

4-(Difluoromethoxy)aniline, 97%

Product Code. 11494237
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494237 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11494237 Supplier Thermo Scientific Alfa Aesar Supplier No. B20349.88
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 22236-10-8 | C7H7F2NO | 159.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 22236-10-8
Density 1.286
Boiling Point 231°C
Flash Point >110°C
Odor Characteristic
Molecular Formula C7H7F2NO
Refractive Index 1.5052
MDL Number MFCD00085005
Quantity 2.5 g
UN Number UN2810
Beilstein 2804462
Synonym 4-difluoromethoxy aniline, 4-difluoromethoxy-phenylamine, p-difluoromethoxyaniline, 4-amino-alpha,alpha-difluoroanisole, 4-difluoromethoxy benzenamine
InChI Key NDEZTSHWEPQVBX-UHFFFAOYSA-N
SMILES NC1=CC=C(OC(F)F)C=C1
IUPAC Name 4-(difluoromethoxy)aniline
Molecular Weight (g/mol) 159.14
PubChem CID 737363
Formula Weight 159.14
Percent Purity 97%
Chemical Name or Material 4-(Difluoromethoxy)aniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.