missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethoxymethyl)-2-methylpyrimidine, 97%, Thermo Scientific™

Product Code. 10089491
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
Product Code. Quantity unitSize
10089491 1 g 1g
10394912 10 g 10g
10168754 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 10089491

Brand: Thermo Scientific Maybridge SPB04845DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175277-33-5
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00085137
InChI Key VDXHJWMLYFVGDL-UHFFFAOYSA-N
PubChem CID 2799572
IUPAC Name 4-(dimethoxymethyl)-2-methylpyrimidine
SMILES COC(OC)C1=CC=NC(C)=N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4-dimethoxymethyl-2-methylpyrimidine, 2-methylpyrimidine-4-carboxaldehyde dimethylacetal, 4-dimethoxymethyl-2-methyl-pyrimidine, acmc-20a0b8, dimethoxy 2-methylpyrimidin-4-yl methane, pyrimidine,4-dimethoxymethyl-2-methyl, pyrimidine, 4-dimethoxymethyl-2-methyl
Formula Weight 168.19
Percent Purity 97%
Chemical Name or Material 4-(Dimethoxymethyl)-2-methylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.