missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylamino-1-naphthoic acid, 98%

Product Code. 11322117
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322117 5 g 5g
11332117 250 mg 250mg
11312117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11322117 Supplier Thermo Scientific Alfa Aesar Supplier No. L07944.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78062-03-0 | C13H13NO2 | 215.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78062-03-0
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.252
MDL Number MFCD00051797
InChI Key LZQKNRRVTOJUHQ-UHFFFAOYSA-N
Synonym 4-dimethylamino-1-naphthoic acid, 4-dimethylamino naphthalene-1-carboxylic acid, 4-dimethylamino-1-naphthalenecarboxylic acid, 4-dimethylamino naphthalic acid, 4-dimethylamino-1-naphthoicacid, 4-dimethylamino-1-naphthoic acid #, 4-dimethylaminonaphthalenecarboxylic acid, 4-dimethylamino naphthalenecarboxylic acid, 4-dimethylamino-naphthalene-1-carboxylic acid
PubChem CID 522695
IUPAC Name 4-(dimethylamino)naphthalene-1-carboxylic acid
SMILES CN(C)C1=CC=C(C2=CC=CC=C21)C(=O)O

Specifications

Melting Point ∽160°C (decomposition)
Quantity 5 g
Beilstein 2723206
Formula Weight 215.25
Percent Purity 98%
Chemical Name or Material 4-Dimethylamino-1-naphthoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.