missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)cinnamic acid, 99%, Thermo Scientific™

Product Code. 10542801 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10542801 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10542801 Supplier Thermo Scientific Acros Supplier No. 204160010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1552-96-1
CAS Min % 98.5
CAS Max % 100.0
Melting Point 225°C to 228°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C11H13NO2
Linear Formula (CH3)2NC6H4CH=CHCO2H
MDL Number MFCD00004397
Quantity 1 g
Beilstein 14, 522
Synonym 4-dimethylamino cinnamic acid, 4-dimethylaminocinnamic acid, p-dimethylamino cinnamic acid, 3-4-dimethylamino phenyl acrylic acid, 4-n,n-dimethylamino cinnamic acid, 2e-3-4-dimethylamino phenyl prop-2-enoic acid, e-3-4-dimethylaminophenyl prop-2-enoic acid, e-3-4-dimethylamino phenyl acrylic acid
InChI Key CQNPVMCASGWEHM-VMPITWQZSA-N
SMILES CN(C)C1=CC=C(C=C1)C=CC(=O)O
IUPAC Name (E)-3-[4-(dimethylamino)phenyl]prop-2-enoic acid
Molecular Weight (g/mol) 191.23
PubChem CID 1540638
Formula Weight 191.23
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)cinnamic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.