missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)piperidine, 97%

Product Code. 11319006
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319006 5 g 5g
11329006 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319006 Supplier Thermo Scientific Alfa Aesar Supplier No. L20176.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 50533-97-6 | C7H16N2 | 128.22 g/mol

4-(Dimethylamino)piperidine is having molecular features associated with polyamine modulation of N-methyl-D-aspartate receptors. It is utilized as a mechanism for reducing the effects of alcohol dependence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Dimethylamino)piperidine is having molecular features associated with polyamine modulation of N-methyl-D-aspartate receptors. It is utilized as a mechanism for reducing the effects of alcohol dependence.

Notes
Incompatible with air, water, oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50533-97-6
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.22
MDL Number MFCD00023144
InChI Key YFJAIURZMRJPDB-UHFFFAOYSA-N
Synonym 4-dimethylamino piperidine, 4-dimethylaminopiperidine, 4-n,n-dimethylamino piperidine, 4-piperidinamine, n,n-dimethyl, 4-dimethylamino-piperidine, dimethyl-piperidin-4-yl-amine, pubchem6749, acmc-1akc4, dimethylpiperidin-4-ylamine
PubChem CID 417391
IUPAC Name N,N-dimethylpiperidin-4-amine
SMILES CN(C)C1CCNCC1

Specifications

Boiling Point 187°C
Refractive Index 1.476
Quantity 5 g
UN Number UN2735
Beilstein 1190
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material 4-(Dimethylamino)piperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.