missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzoic acid, 98%

Product Code. 10636302 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10636302 100 g Plastic bottle 100g
10294090 25 g Plastic bottle 25g
10095943 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10636302 Supplier Thermo Scientific Acros Supplier No. 115761000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 619-84-1 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-84-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00002537
InChI Key YDIYEOMDOWUDTJ-UHFFFAOYSA-N
Synonym 4-dimethylamino benzoic acid, benzoic acid, 4-dimethylamino, benzoic acid, p-dimethylamino, p-dimethylaminobenzoic acid, n,n-dimethyl-p-aminobenzoic acid, n,n-dimethyl-4-aminobenzoic acid, p-n,n-dimethylamino benzoic acid, p-dimethylamino benzoic acid, unii-d1ma908ev0, 4-n,n-dimethylamino benzoic acid
PubChem CID 12092
IUPAC Name 4-(dimethylamino)benzoic acid
SMILES CN(C)C1=CC=C(C=C1)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 238°C to 242°C
Color Beige-White to Gray-Green
Density 170g/mL
Flash Point 170°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula (CH3)2NC6H4CO2H
Quantity 100 g
Beilstein 14, 426
Merck Index 15, 3254
Solubility Information Solubility in water: .. Other solubilities: soluble in hcl and koh solutions,soluble in alcohol,sparingly soluble in ether,practically insoluble in acetic acid
Formula Weight 165.19
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Dimethylaminobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.