missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzophenone, 98%

Product Code. 11484307
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484307 100 g 100g
11464307 5 g 5g
11474307 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484307 Supplier Thermo Scientific Alfa Aesar Supplier No. B20389.22
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 530-44-9 | C15H15NO | 225.291 g/mol

4-(Dimethylamino)benzophenone is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Dimethylamino)benzophenone is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-44-9
Molecular Formula C15H15NO
Molecular Weight (g/mol) 225.291
MDL Number MFCD00008311
InChI Key BEUGBYXJXMVRFO-UHFFFAOYSA-N
Synonym 4-dimethylamino benzophenone, 4-dimethylaminobenzophenone, p-dimethylaminobenzophenone, p-benzoyl-n,n-dimethylaniline, 4-n,n-dimethylaminobenzophenone, p-dimethylamino benzophenone, 4-dimethylamino phenyl phenyl methanone, 4-benzoyl-n,n-dimethylaniline, methanone, 4-dimethylamino phenyl phenyl, benzophenone, 4-dimethylamino
PubChem CID 10737
IUPAC Name [4-(dimethylamino)phenyl]-phenylmethanone
SMILES CN(C)C1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 88°C to 90°C
Quantity 100 g
Merck Index 14,3233
Solubility Information Insoluble in water.
Formula Weight 225.29
Percent Purity 98%
Chemical Name or Material 4-Dimethylaminobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.