missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminocinnamic acid, 97+%

Product Code. 11487124
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487124 1 g 1g
11497124 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11487124 Supplier Thermo Scientific Alfa Aesar Supplier No. B21549.03
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1552-96-1 | C11H13NO2 | 191.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1552-96-1
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00004397
InChI Key CQNPVMCASGWEHM-VMPITWQZSA-N
Synonym 4-dimethylamino cinnamic acid, 4-dimethylaminocinnamic acid, p-dimethylamino cinnamic acid, 3-4-dimethylamino phenyl acrylic acid, 4-n,n-dimethylamino cinnamic acid, 2e-3-4-dimethylamino phenyl prop-2-enoic acid, e-3-4-dimethylaminophenyl prop-2-enoic acid, e-3-4-dimethylamino phenyl acrylic acid
PubChem CID 1540638
IUPAC Name (E)-3-[4-(dimethylamino)phenyl]prop-2-enoic acid
SMILES CN(C)C1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point ∼222°C (decomposition)
Quantity 1 g
Beilstein 1368533
Formula Weight 191.23
Percent Purity 99%
Chemical Name or Material 4-Dimethylaminocinnamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.