missing translation for 'onlineSavingsMsg'
Learn More

4-[(Dipropylamino)sulfonyl]benzoic acid, 97%, Maybridge™

Product Code. 10621503
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10621503 10 g 10g
10503145 25 g 25g
11326316 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10621503 Supplier Thermo Scientific Maybridge Supplier No. SB00915EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 57-66-9
Molecular Formula C13H19NO4S
Molecular Weight (g/mol) 285.36
MDL Number MFCD00038402
InChI Key DBABZHXKTCFAPX-UHFFFAOYSA-N
Synonym probenecid, 4-dipropylsulfamoyl benzoic acid, probenecid acid, probecid, benecid, benemid, benuryl, probalan, probenemid, probexin
PubChem CID 4911
ChEBI CHEBI:8426
IUPAC Name 4-(dipropylsulfamoyl)benzoic acid
SMILES CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula [CH3(CH2)2]2NSO2C6H4CO2H
Quantity 10 g
Formula Weight 285.36
Percent Purity 97%
Chemical Name or Material 4-[(Dipropylamino)sulfonyl]benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.