missing translation for 'onlineSavingsMsg'
Learn More

4-Epianhydrochlortetracycline Hydrochloride (>90%), TRC

Product Code. 30132461
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132461 5 mg 5mg
30133558 1 mg 1mg
30006575 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132461 Supplier Toronto Research Chemicals Supplier No. TRCE5780605MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158018-53-2
Molecular Formula C22 H21 Cl N2 O7 . Cl H
Molecular Weight (g/mol) 497.33
Synonym [4R-(4alpha,4abeta,12abeta)]-7-Chloro-4-(dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide Monohydrochloride;
IUPAC Name (4R,4aS,12aS)-7-chloro-4-(dimethylamino)-3,10,12,12a-tetrahydroxy-6-methyl-1,11-dioxo-4a,5-dihydro-4H-tetracene-2-carboxamide;hydrochloride
SMILES Cl.CN(C)[C@@H]1[C@@H]2CC3=C(C)c4c(Cl)ccc(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H21ClN2O7.ClH/c1-7-8-6-9-16(25(2)3)18(28)15(21(24)31)20(30)22(9,32)19(29)13(8)17(27)14-11(26)5-4-10(23)12(7)14;/h4-5,9,16,26,28-29,32H,6H2,1-3H3,(H2,24,31);1H/t9-,16+,22-;/m0./s1
Formula Weight 496.0804
Percent Purity >95%
Chemical Name or Material 4-Epianhydrochlortetracycline hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.