missing translation for 'onlineSavingsMsg'
Learn More

4-Epianhydrochlortetracycline hydrochloride, can be used as secondary standard, Thermo Scientific Chemicals

Product Code. 10073320 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10073320 100 mg Glass bottle 100mg
10308960 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10073320 Supplier Thermo Scientific Acros Supplier No. 268221000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158018-53-2
Molecular Formula C22H21ClN2O7·HCl
Molecular Weight (g/mol) 497.33
MDL Number MFCD00143313
InChI Key ISGAAFMBTIWTEU-PXAZKYFKSA-N
Synonym 4-epianhydrochlortetracycline hydrochloride, 4-epianhydrochlortetracycline hcl, epianhydrochlortetracycline hydrochloride, 2z,4r,4as,12as-2-amino hydroxy methylidene-7-chloro-4-dimethylamino-10,11,12a-trihydroxy-6-methyl-4a,5-dihydro-4h-tetracene-1,3,12-trione hydrochloride, 4r,4as,12ar-7-chloro-4-dimethylamino-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-3,4,4a,5,12,12a-hexahydrotetracene-2-carboxamide-hydrogen chloride 1/1, 4r,4as,12as-7-chloro-4-dimethylamino-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-4a,5-dihydro-4h-tetracene-2-carboxamide hydrochloride, 4r-4, a,4a, a,12a, a-7-chloro-4-dimethylamino-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide monohydrochloride
PubChem CID 54710413
IUPAC Name (4R,4aS,12aR)-7-chloro-4-(dimethylamino)-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-4a,5-dihydro-4H-tetracene-2-carboxamide;hydrochloride
SMILES CC1=C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C14)Cl)O)O)O)O)C(=O)N)N(C)C.Cl

Specifications

Melting Point 222.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 80% min. (as is,calculated)
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: 10g/L. Other solubilities: 29g/L methanol
Formula Weight 497.33
Percent Purity ≥97.0% Total Base (on anhydrous substance), ≥80 wt% (as is, calculated) (HPLC)
Physical Form Powder
Chemical Name or Material 4-Epianhydrochlortetracycline hydrochloride, Can be used as secondary standard
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Epianhydrochlortetracycline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.