missing translation for 'onlineSavingsMsg'
Learn More

4-Epianhydrotetracycline Hydrochloride Hydrate (>85%), TRC

Product Code. 30377836
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30377836 10 mg
30369624 25 mg
30377539 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377836 Supplier Toronto Research Chemicals Supplier No. TRCE57807010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C22H22N2O7.HCl.H2O
Molecular Weight (g/mol) 480.9
Synonym (4R,4aS,12aS)-4-(Dimethylamino)-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide Hydrochloride
IUPAC Name (4R,4aS,12aS)-4-(dimethylamino)-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-1,4,4a,5,12,12a-hexahydrotetracene-2-carboxamide hydrochloride hydrate
SMILES O[C@@]12[C@]([H])([C@@H](N(C)C)C(O)=C(C(N)=O)C2=O)CC3=C(C4=CC=CC(O)=C4C(O)=C3C1=O)C.Cl.O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H22N2O7.ClH.H2O/c1-8-9-5-4-6-12(25)13(9)17(26)14-10(8)7-11-16(24(2)3)18(27)15(21(23)30)20(29)22(11,31)19(14)28;;/h4-6,11,16,25-27,31H,7H2,1-3H3,(H2,23,30);1H;1H2/t11-,16+,22-;;/m0../s1
Formula Weight 480.13
Chemical Name or Material 4-Epianhydrotetracycline Hydrochloride Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.