missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzonitrile, 98%

Product Code. 11438667
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438667 100 g 100g
11418667 5 g 5g
11428667 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438667 Supplier Thermo Scientific Alfa Aesar Supplier No. A19806.22
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 25117-74-2 | C9H9NO | 147.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25117-74-2
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00001819
InChI Key PJRLUGQMEZZDIY-UHFFFAOYSA-N
Synonym benzonitrile, 4-ethoxy, p-ethoxycyanobenzene, p-ethoxybenzonitrile, 4-ethoxybenzoic acid nitrile, benzonitrile, p-ethoxy, 4-ethoxybenzenecarbonitrile, 4-ethoxy-benzonitrile, 4-ethyloxybenzonitrile, acmc-1cis8
PubChem CID 141176
IUPAC Name 4-ethoxybenzonitrile
SMILES CCOC1=CC=C(C=C1)C#N

Specifications

Melting Point 60°C to 63°C
Boiling Point 258°C
Quantity 100 g
UN Number UN3439
Beilstein 2613602
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material 4-Ethoxybenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.