missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyl-2-methoxyphenol, 98%

Product Code. 11462357
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462357 5 g 5g
11472357 25 g 25g
11482357 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462357 Supplier Thermo Scientific Alfa Aesar Supplier No. A14239.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 2785-89-9 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 2785-89-9
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00038714
InChI Key CHWNEIVBYREQRF-UHFFFAOYSA-N
Synonym 4-ethylguaiacol, p-ethylguaiacol, 2-methoxy-4-ethylphenol, phenol, 4-ethyl-2-methoxy, homocresol, guaiacyl ethane, 4-ethyl-2-methoxy-phenol, 4-hydroxy-3-methoxy ethylbenzene, unii-c9nfd83bj5, guaiacol, 4-ethyl
PubChem CID 62465
IUPAC Name 4-ethyl-2-methoxyphenol
SMILES CCC1=CC=C(O)C(OC)=C1

Specifikationer

Melting Point 15°C
Density 1.064
Boiling Point 230°C
Flash Point 107°C (225°F)
Refractive Index 1.528
Quantity 5 g
Beilstein 2045329
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 4-Ethyl-2-methoxyphenol

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.