missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylanisole, 98+%

Product Code. 11418573
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418573 50 g 50g
11408573 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11418573 Supplier Thermo Scientific Alfa Aesar Supplier No. A10391.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1515-95-3 | C9H12O | 136.194 g/mol

It can be used the synthesis of 1-Methoxy-4-ethyl-1,4-cyclohexadiene. It is also applied in the ligand-assisted, copper-catalyzed enantioselective benzylic amination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used the synthesis of 1-Methoxy-4-ethyl-1,4-cyclohexadiene. It is also applied in the ligand-assisted, copper-catalyzed enantioselective benzylic amination.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1515-95-3
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00015176
InChI Key HDNRAPAFJLXKBV-UHFFFAOYSA-N
Synonym 4-ethylanisole, p-ethylanisole, benzene, 1-ethyl-4-methoxy, anisole, p-ethyl, p-ethylanisol, unii-i71p5qg20s, 4-ethyl-1-methoxybenzene, 4-ethyl anisole, 4-ethylmethoxybenzene, acmc-1btzv
PubChem CID 73690
IUPAC Name 1-ethyl-4-methoxybenzene
SMILES CCC1=CC=C(C=C1)OC

Specifications

Density 0.962
Boiling Point 195°C
Flash Point 79°C (174°F)
Refractive Index 1.5085
Quantity 50 g
Beilstein 774365
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 136.19
Percent Purity ≥98%
Chemical Name or Material 4-Ethylanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.