missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzoic acid, 99%

Product Code. 11463933
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11463933

Brand: Thermo Scientific Alfa Aesar A10235.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 619-64-7 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-64-7
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002570
InChI Key ZQVKTHRQIXSMGY-UHFFFAOYSA-N
Synonym benzoic acid, 4-ethyl, p-ethylbenzoic acid, 4-ethyl benzoic acid, 4-ethyl-benzoic acid, unii-9tkm8p0g6r, 9tkm8p0g6r, p-ethylbenzoate, 4-ethyl benzoate, 4-ethylbenzoicacid, p-ethylbenzoic acid;
PubChem CID 12086
IUPAC Name 4-ethylbenzoic acid
SMILES CCC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 111°C to 114°C
Quantity 250 g
Beilstein 2041840
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material 4-Ethylbenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.